Orders LinkContactUsLinkComponent
Plastic Boxes, Safety Closure | Avantor
Plastic Boxes, Safety Closure
Supplier:   MULTIROIR
Plastic Boxes, Safety Closure
Supplier:  MULTIROIR
Certificates
Plastic Boxes, Safety Closure
Plastic Boxes, Safety Closure
6 Options Available Below
Product Details & Documents
The plastic boxes with secure lid allow the storage of your liquid and solid products.
  • Safety closure
  • 100% liquid-tight
  • Highly impact-resistant PP material
  • Dishwasher safe. Microwave safe
  • Food contact
  • Material compatible with very low temperatures
  • Standard colour: Transparent
They offer a perfect seal and a temper-evident closure, especially for liquid products.
Dokumente
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 0,90 L
    128×128×89 mm
    Log in for availability
    Log in for availability
    1 * 180 ST
    1 * 180 ST - 390,00 €
    390,00 €
    1 * 180 ST
       390,00 €
  • 1,10 L
    128×128×118 mm
    Log in for availability
    Log in for availability
    1 * 150 ST
    1 * 150 ST - 356,00 €
    356,00 €
    1 * 150 ST
       356,00 €
  • 2 L
    192×192×77 mm
    Log in for availability
    Log in for availability
    1 * 78 ST
    1 * 78 ST - 312,00 €
    312,00 €
    1 * 78 ST
       312,00 €
  • 3 L
    192×192×112 mm
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 325,00 €
    325,00 €
    1 * 72 ST
       325,00 €
  • 4 L
    192×192×150 mm
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 250,00 €
    250,00 €
    1 * 48 ST
       250,00 €
  • 5 L
    192×192×191 mm
    Log in for availability
    Log in for availability
    1 * 36 ST
    1 * 36 ST - 263,00 €
    263,00 €
    1 * 36 ST
       263,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.