Order Entry
Austria
ContactUsLinkComponent
Calcium chloride solution 0.5 M for molecular biology
Calcium chloride solution 0.5 M for molecular biology
Catalog # AVIVOORA00231
CAS Number:  
Calcium chloride solution 0.5 M for molecular biology
Catalog # AVIVOORA00231
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903