Order Entry
Austria
ContactUsLinkComponent
L(+)-Glutamine, powder cell culture reagent, Corning®
L(+)-Glutamine, powder cell culture reagent, Corning®
Supplier:  Corning
Certificates
undefined
L(+)-Glutamine, powder cell culture reagent, Corning®
Supplier:  Corning
Certificates

Specification Test Results

  • Appearance:
    White crystals or crystalline powder
  • Identification :
    Passes test
  • Assay :
    98.5% - 101.5% (calculated on dried basis)
  • Residue on Ignition:
    NMT 0.3%
  • Chloride :
    NMT 0.05%
  • Sulfate:
    NMT 0.03%
  • Iron:
    NMT 30 ppm
  • Heavy Metals :
    NMT 15 ppm
  • Related Compounds:
    NMT 0.5%
  • Specific Rotation at 20 °C:
    +6.3 ° to 7.3 °
  • Loss on Drying:
    NMT 0.3%
  • Identification:
    Passes test
  • Assay:
    98.5%-10_1.5% (on dried basis)
  • Lead:
    NMT 5mg/kg
  • Specific Rotation at 20°C:
    +6.3 ° to 7.3 °

Specification Test Results

Frequently Bought Together