Order Entry
Austria
ContactUsLinkComponent
 
Anti-PTPRC Rat Antibody (APC (Allophycocyanin)-Cy7®) [clone: LTF-2]
  25-4031-U100
undefined
Anti-PTPRC Rat Antibody (APC (Allophycocyanin)-Cy7®) [clone: LTF-2]
  25-4031-U100

 

  • Antibody type:
    Control
  • Antigen name:
    Protein Tyrosine Phosphatase Receptor Type C
  • Antigen symbol:
    PTPRC
  • Clone:
    LTF-2
  • Conjugation:
    APC (Allophycocyanin)-Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Neutralisation:
    Yes
  • Antigen synonyms:
    CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Storage temperature:
    Store Undiluted at 2 to 8 °C; Protect from Light; Do Not Freeze.
  • Tested applications:
    Flow Cytometry
  • Pk:
    100 µG

 

 

Type: Control
Antigen: PTPRC
Clonality:
Clone: LTF-2
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: 0