Order Entry
Austria
ContactUsLinkComponent
Rat IgG1 Isotype Control [clone: HRPN] (FITC (Fluorescein Isothiocyanate))
Rat IgG1 Isotype Control [clone: HRPN] (FITC (Fluorescein Isothiocyanate))
  35-4301-U100
undefined
Rat IgG1 Isotype Control [clone: HRPN] (FITC (Fluorescein Isothiocyanate))
  35-4301-U100

 

  • Antibody type:
    Control
  • Clone:
    HRPN
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG1
  • Storage temperature:
    Store Undiluted at 2 to 8 °C; Protect from Light; Do Not Freeze.
  • Tested applications:
    Flow Cytometry
  • Pk:
    100 µG

 

 

The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Type: Control
Antigen: HRPN
Clonality:
Clone: HRPN
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: