About this item
The Solus™ CCS safety glasses feature a rimless design with lime green temples and a corded control system (CCS) for attaching corded earplugs. The polycarbonate lens features a Scotchgard™ anti-fog and anti-scratch coating on both sides for increased durability and improved vision.
- Modern safety glasses range keeps the eyewear and corded earplugs attached and ready to use
- Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties (available on selected models)
- Suitable for use with all non-textile 3M™ corded earplugs
- Available with a removeable foam gasket
- Frame marking: 3M EN 166 FT CE
For users who are intermittently exposed to noise, use 3M™ Solus™ CCS safety glasses. These rimless safety glasses feature lime green temples that have a corded control system (CCS) which allows users to keep certain E-A-R™ corded earplugs attached and ready for use at all times. The unique temple design holds the cord of the earplugs to keep them securely in place and untangled. When the corded earplugs are not in use, the CCS feature allows them to act as a lanyard for carrying the protective eyewear around the neck.
The polycarbonate lens features Scotchgard™ anti-fog and anti-scratch coating for superior anti-fog protection and increased durability that resists fogging for longer than traditional anti-fog coatings, even after up to 25 washings with water.