Orders LinkContactUsLinkComponent
Conductive tips, MBP
Conductive tips, MBP
Tecan style art tips, 1000 µl, +, Black
Catalog # MLBP904-013
   Low Manufacturing Impact
Conductive tips, MBP
Catalog # MLBP904-013
   Low Manufacturing Impact
Conductive tips, MBP
Conductive tips, MBP
Tecan style art tips, 1000 µl, +, Black
Price
1 090,00 €
Log in for availability
1 * 4.800 ST
Product Details & Documents
Filtered, conductive tip style, liquid sensing, Econopacks.
  • For use with: Tecan™ Freedom EVO™ LiHa Head, Tecan™ Genesis™
  • Ideal for automated liquid handling systems
  • Non sterile, polypropylene
Perkin-Elmer is a registered trademark of Perkin-Elmer Corporation. ART is a registered trademark of MBP and is protected by U.S. Patent # 5,156,811. EconoPak is a trademark of Molecular BioProducts. Tecan is a registered trademark of Tecan AG. Cavro is a registered trademark of Scientific Instruments, Inc. MultiPROBE and Packard are registered trademarks of Packard Instrument Company, Inc. Thermo Orion is a registered trademark of Thermo Orion Inc.
Specifications
  • Volume:
    1000 µl
  • Tip style:
    Tecan style art tips
  • Tip colour:
    Black
  • Filter:
    +
  • Cat. No.:
    MLBP904-013
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact