Anti-STING mouse monoclonal antibody [clone: 1B10]
CAYM25923-100
- Antibody type:Primary
- Antigen symbol:STING
- Clonality:Monoclonal
- Clone:1B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminus
- Form:Liquid
- Amino acid number:351-379
- UniProtKB:Q86WV6
- Storage buffer:PBS, pH 7,2, with 50% glycerol and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:Protein G-purified monoclonal antibody
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: STING (C-Term) Monoclonal Antibody (Clone 1B10)
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope: C-terminus
Host: Mouse
Isotype: IgG1
Reactivity: Human