Anti-ALIX rabbit polyclonal antibody
Catalog # NOVUNBP1-49701
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Apoptosis-Linked Gene 2-Interacting Protein X
- Antigen symbol:Alix
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:10015
- Antigen synonyms:ALG-2 interacting protein X|programmed cell death 6 interacting protein|ALIX|dopamine receptor interacting protein 4|ALG-2-interacting protein 1|apoptosis-linked gene 2-interacting protein X|alinx|ALG-2 interacting protein 1|HP95|programmed cell death 6-interacting protein|PDCD6-interacting protein|KIAA1375|AIP1DRIP4|MGC17003
- Storage buffer:TBS and 0,1% BSA
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:0,2 mg/ml
- Immunogen:The immunogen for this product maps to a region between residue 818 and 868 of human Apoptosis-Linked Gene 2-Interacting Protein X using the numbering given in entry NP_037506.2 (GeneID 10015).
- Purification:Immunogen affinity purified
- Pk:0,1 mL
Specifications
About this item
The Alix Antibody from Novus Biologicals is a rabbit polyclonal antibody to Alix. This antibody reacts with human. The Alix Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation.
Type: Primary
Antigen: Alix
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human