Order Entry
Austria
ContactUsLinkComponent
Human recombinant p62/SQSTM1 (from E. coli)
Human recombinant p62/SQSTM1 (from E. coli)
Catalog # NOVUNBP1-44490
Supplier:  Novus Biologicals
CAS Number:  
undefined
Human recombinant p62/SQSTM1 (from E. coli)
Catalog # NOVUNBP1-44490
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    0,1 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
  • Gene ID:
    8878
  • Protein synonyms:
    sequestosome 1|Paget disease of bone 3|ORCA|OSIL|p60PDB3|oxidative stress induced like|EBI3-associated protein of 60 kDa|EBIAP|A170|Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa|ZIP3|p62B|sequestosome-1|p62|EBI3-associated protein p60|Ubiquitin-binding protein p62|phosphotyrosine independent ligand for the Lck SH2 domain p62
  • Protein/peptide name:
    p62/SQSTM1
  • Purity:
    >90%, by SDS-PAGE
  • Concentration:
    1 mg/ml
  • Tested applications:
    ELISA

Specifications

About this item

The Recombinant Human p62/SQSTM1 Protein from Novus Biologicals is derived from E. coli. The Recombinant Human p62/SQSTM1 Protein has been validated for the following applications: SDS-Page.