Mouse monoclonal anti-Smith antigen antibody (Y12)
Catalog # NOVUNB600-546
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Smith Antigen
- Antigen symbol:Smith Antigen
- Clonality:Monoclonal
- Clone:Y12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG3 kappa
- Reactivity:Drosophila,Pig,Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:Smith antigen
- Storage buffer:PBS (pH 7,4) and 0,2% BSA
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen is generated from human Smith Antigen.
- Tested applications:Immunomicroscopy
- Purification:Protein G purified
- Pk:500 µl
Specifications
About this item
The Smith Antigen Antibody (Y12) from Novus Biologicals is a mouse monoclonal antibody to Smith Antigen. This antibody reacts with human, mouse, rat, porcine, drosophila, rabbit. The Smith Antigen Antibody (Y12) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunomicroscopy, Immunofluorescence.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:100
Type: Primary
Antigen: Smith Antigen
Clonality: Monoclonal
Clone: Y12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human, Mouse, Rat, Porcine, Drosophila, Rabbit