Software for SCIEX QTRAP 4500, Triple Quad 3500 and Triple Quad 4500 LC-MS/MS systems
Supplier: AB SCIEX
Certificates
Software for SCIEX QTRAP 4500, Triple Quad 3500 and Triple Quad 4500 LC-MS/MS systems
Supplier: AB SCIEX
About this item
Frequently Bought Together
VWR® SAS Super Isolator Air Samplers
Supplier: VWR Collection
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
VWR® VisiCam® Plus, Digital USB Cameras
Supplier: VWR Collection
CV Glassy carbon for ElectraSyn 2.0 1 * 1 items
Supplier: IKA
Nuts and ferrules, Super Flangeless™, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Transfer tanks, Transphor™
Supplier: Cytiva
OpenLab CDS Software
Supplier: VWR Collection
VWR®, Thermal Cyclers
Supplier: VWR Collection
pH meter, SevenDirect SD20
Supplier: Mettler - Toledo
UV bench lamps, XX-series
Supplier: UVP ULTRA VIOLET PRODUCTS
Storage phosphor screens and cassettes
Supplier: Cytiva
Accessories for multiparameter photometer, Water-ID® The PrimeLab 2.0
Supplier: WATER ID PRIMELAB