Mouse IgG2b Isotype Control [clone: MPC-11]
Catalog # 40-4732-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Antigen name:Mitochondrial pyruvate carrier 11
- Clone:MPC-11
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:
- Environmentally Preferable:
- Neutralisation:Yes
- Storage temperature:Store undiluted at 2...8 °C, protect from light, do not freeze.
- Tested applications:Functional Assay
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.