Anti-Sars Spike Protein Rabbit polyclonal antibody
Catalog # NOVUNB100-56578SS
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Sars Spike Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:1489668
- Antigen synonyms:Spike glycoprotein|VGL2|E2|S|Peplomer protein|Human coronavirus spike glycoprotein|Severe acute respiratory syndrome virus spike glycoprotein|S glycoprotein|Severe acute respiratory syndrome spike glycoprotein|E2 glycoprotein
- Storage buffer:PBS containing 0.05% BSA
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:0,5 mg/ml
- Immunogen:The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 1124-1140 of the putative SARS Spike glycoprotein (Genbank accession no. NP_828851).
- Purification:Protein G purified
- Pk:25 µl
Specifications
About this item
The Sars Spike Protein Antibody from Novus Biologicals is a rabbit polyclonal antibody to Sars Spike Protein. This antibody reacts with human. The Sars Spike Protein Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Sars Spike Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human