Order Entry
Austria
ContactUsLinkComponent
Rabbit Polyclonal anti-160kDa Neurofilament Medium antibody
Rabbit Polyclonal anti-160kDa Neurofilament Medium antibody
Catalog # NOVUNB300-133
Supplier:  Novus Biologicals
Rabbit Polyclonal anti-160kDa Neurofilament Medium antibody
Catalog # NOVUNB300-133
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    160 kDa Neurofilament Medium
  • Antigen symbol:
    NF-M
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Pig,
    Birds,
    Human,
    Bovine,
    Rat,
    Mouse,
    Cat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    4741
  • Antigen synonyms:
    Neurofilament 3|medium polypeptide 150kDa|NEF3neurofilament medium polypeptide|neurofilament|NFMNeurofilament triplet M protein|neurofilament-3 (150 kD medium)|medium polypeptide|NF-M160 kDa neurofilament protein
  • Storage buffer:
    Whole antisera
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    Recombinant rat Neurofilament Medium fusion protein corresponding to the C-terminus [UniProt# P12839]
  • Purification:
    Unpurified
  • Pk:
    0,1 mL

Specifications

About this item

The NF-M Antibody from Novus Biologicals is a rabbit polyclonal antibody to NF-M. This antibody reacts with human, mouse, rat, porcine, avian, bovine, feline. The NF-M Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000

Type: Primary
Antigen: NF-M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Avian, Bovine, Feline