Order Entry
Austria
ContactUsLinkComponent
Sodium dihydrogen phosphate dihydrate for HPLC
Sodium dihydrogen phosphate dihydrate for HPLC
  ACRO446222500
 :  
undefined
Sodium dihydrogen phosphate dihydrate for HPLC
  ACRO446222500
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Appearance (Color):
    White
  • Appearance (Form):
    Crystals
  • UV at 254 nm A (1 M in water):
    =<0.05
  • pH:
    4.0 to 4.5
  • Titration Acid:
    98.5 to 101.5 %
  • Water:
    =<24 %

 

 

 :
 :156.01 g/mol
 :12,08806
 :60 °C
 :Ambient
 :231-449-2
 :01-2119489796-13
 :13472-35-0
 :NaH₂PO₄·2H₂O
 :MFCD00149209