Order Entry
Austria
ContactUsLinkComponent
Iodine alcoholic solution Reag.Ph.Eur. 1045804
Iodine alcoholic solution Reag.Ph.Eur. 1045804
Catalog # 84119.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Iodine alcoholic solution Reag.Ph.Eur. 1045804
Catalog # 84119.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

 :
 :253.81 g/mol
Storage Temperature:Ambient
UN:1170
ADR:3,II
 :GHS02
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355