Sodium hydroxide 32% in aqueous solution for biopharmaceutical production
: Caustic soda
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:Ambient
:215-185-5
:1824
:8,II
:GHS05
- Environmentally Preferable:
- Assay (calculated as NaOH) 31.8 - 32.2 %:31.8 - 32.2 %
- Appearance:Passes test
- CO3 (as Na₂CO₃):≤ 1 %
- Heavy metals (as Pb):≤ 5 ppm
- Cl (Chloride):≤ 20 ppm
- SO₄ (Sulphate):≤ 30 ppm
- Fe (Iron):≤ 3 ppm
- Residual metal catalysts:Unlikely to be present
- Residual solvents (ICH Q3C):Excl.by manufact. process
- Endotoxin:≤ 2.0 EU/ml
Frequently Bought Together