Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, Cyanide 1000 mg/L Standard for ion chromatography in 0.1% KOH, Certified Reference Material
VWR® ARISTAR®, Cyanide 1000 mg/L Standard for ion chromatography in 0.1% KOH, Certified Reference Material
Catalog # 85784.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Cyanide 1000 mg/L Standard for ion chromatography in 0.1% KOH, Certified Reference Material
Catalog # 85784.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Matrix:
    0.1% KOH
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Cyanide standard solution
  • Storage temperature:
    2°C - 8°C

Specifications

About this item

This certified reference material is produced using a high purity starting material, acid from sub-boiling and 18 MOhm deionized water and filtered through a 0.2 micron filter.

Certified value and uncertainty (mg/l): 1000.0 ± 5.0

Certifications: CRM ISO 17034 and ISO 17025. Traceable to NIST.