ContactUsLinkComponent
Powered air respirator visor frame assemblies, Versaflo™
Powered air respirator visor frame assemblies, Versaflo™
Catalog # MMMAM-961
Supplier:  3M
undefined
Powered air respirator visor frame assemblies, Versaflo™
Catalog # MMMAM-961
Supplier:  3M
Specifications
  • Description:
    Visor frame assemblies
  • Pk:
    1
Specifications
About this item
The Versaflo™ Over-Visor Frame M-961 is a carrier frame designed to fit Versaflo™ Over-Visor M-967 and Versaflo™ radiant heat covers for Versaflo™ M-Series Helmets.
  • Carrier frame to fit over-visor and radiant heat covers to M-Series helmets
  • Designed specifically for use with M-Series helmets
This frame is made from aluminium.