Orders LinkContactUsLinkComponent
Compartment Boxes
Supplier:   MULTIROIR
Compartment Boxes
Supplier:  MULTIROIR
Certificates
Compartment Boxes
Compartment Boxes
10 Options Available Below
Product Details & Documents
Compartment boxes are suitable for storage, transport, and presentation of small parts. All models are provided with a perfectly transparent cover in order to showcase the contents inside.
  • High transparency lid offers optimum visibility of contents
  • 100% recyclable virgin material
  • Box material: Butadiene styrene copolymer
  • Inserts material: Polystyrene
  • Transparent cover with secure closure by simple pressure
  • Perfect stacking stability
The boxes are equipped with white removable inserts. They are made from high quality materials that are strong and durable. The thickness of the material, the careful design, and the quality of design make them excellent presentation boxes for professionals.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 216-2487
    15-compartment box, with 45×45 mm compartment sizes
    15-compartment box, with 45×45 mm compartment sizes
    285×177×25 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 226,00 €
    226,00 €
    1 * 10 ST
       226,00 €
  • 216-2488
    31-compartment box, with 150×45 / 31×26 mm compartment sizes
    31-compartment box, with 150×45 / 31×26 mm compartment sizes
    285×177×25 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 226,00 €
    226,00 €
    1 * 10 ST
       226,00 €
  • 216-2489
    54-compartment box, with 25×20 mm compartment sizes
    54-compartment box, with 25×20 mm compartment sizes
    285×177×25 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 226,00 €
    226,00 €
    1 * 10 ST
       226,00 €
  • 216-2480
    5-compartment box, with 151×65 / 60×60 / 194×85 mm compartment sizes
    5-compartment box, with 151×65 / 60×60 / 194×85 mm compartment sizes
    285×177×45 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 206,00 €
    206,00 €
    1 * 8 ST
       206,00 €
  • 216-2481
    6-compartment box, with 85×73 mm compartment sizes
    6-compartment box, with 85×73 mm compartment sizes
    285×177×45 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 206,00 €
    206,00 €
    1 * 8 ST
       206,00 €
  • 216-2482
    12-compartment box, with 62×47 mm compartment sizes
    12-compartment box, with 62×47 mm compartment sizes
    285×177×45 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 206,00 €
    206,00 €
    1 * 8 ST
       206,00 €
  • 216-2483
    15-compartment box, with 47×47 mm compartment sizes
    15-compartment box, with 47×47 mm compartment sizes
    285×177×45 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 206,00 €
    206,00 €
    1 * 8 ST
       206,00 €
  • 216-2484
    28-compartment box, with 35×35 mm compartment sizes
    28-compartment box, with 35×35 mm compartment sizes
    285×177×45 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 206,00 €
    206,00 €
    1 * 8 ST
       206,00 €
  • 216-2485
    31-compartment box, with 31×27 / 45×150 mm compartment sizes
    31-compartment box, with 31×27 / 45×150 mm compartment sizes
    285×177×45 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 206,00 €
    206,00 €
    1 * 8 ST
       206,00 €
  • 216-2486
    54-compartment box, with 24×22 mm compartment sizes
    54-compartment box, with 24×22 mm compartment sizes
    285×177×45 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 206,00 €
    206,00 €
    1 * 8 ST
       206,00 €