Orders LinkContactUsLinkComponent
ProBox storage boxes
ProBox storage boxes
Supplier:   MULTIROIR
ProBox storage boxes
Supplier:  MULTIROIR
Certificates
ProBox storage boxes
ProBox storage boxes
7 Options Available Below
Product Details & Documents
These storages boxes are a premium organising system developed with a vision of functionality, high quality, and a clear Scandinavian design.
  • Made of translucent material PP (polypropylene)
  • Stackable
  • Guaranteed food-safe
  • Hygienically flawless and easy to clean
  • Ergonomic handles
  • Suitable for –40 to 120 °C storage
  • 100% recyclable
The volume of the boxes ranges between 3 and 53 L, which makes the range suitable for a wide range of storage needs. All boxes are modular and stackable for optimal space-saving storage. The recess and furrows in the lid prevents them from shifting when stacked. The boxes are translucent and the lid is easily clicked on with two wide and strong grey clips.
Documents
Product Family Options
Product Information
  • Capacity
    W×D×H
    Material
    Availability
    Price
  • 3 L
    Exterior: 290×195×98 mm
    Interior: 255×135×80 mm
    PP
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 54,70 €
    54,70 €
    1 * 5 ST
       54,70 €
  • 6 L
    Exterior: 290×195×185 mm
    Interior: 255×135×170 mm
    PP
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 64,70 €
    64,70 €
    1 * 5 ST
       64,70 €
  • 8 L
    Exterior: 390×290×108 mm
    Interior: 330×250×90 mm
    PP
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 87,00 €
    87,00 €
    1 * 5 ST
       87,00 €
  • 14 L
    Exterior: 390×290×185 mm
    Interior: 330×250×170 mm
    PP
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 87,00 €
    87,00 €
    1 * 5 ST
       87,00 €
  • 21 L
    Exterior: 510×390×169 mm
    Interior: 420×330×150 mm
    PP
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 150,00 €
    150,00 €
    1 * 5 ST
       150,00 €
  • 32 L
    Exterior: 510×390×258 mm
    Interior: 420×330×235 mm
    PP
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 167,00 €
    167,00 €
    1 * 5 ST
       167,00 €
  • 53 L
    Exterior: 510×390×435 mm
    Interior: 420×330×415 mm
    PP
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 246,00 €
    246,00 €
    1 * 5 ST
       246,00 €