Order Entry
Austria
ContactUsLinkComponent
Bovine calf serum, 1 L, iron-fortified, United States origin, Corning®
Bovine calf serum, 1 L, iron-fortified, United States origin, Corning®
Catalog # MDTC35-053-CM
Supplier:  Corning
Bovine calf serum, 1 L, iron-fortified, United States origin, Corning®
Catalog # MDTC35-053-CM
Supplier:  Corning

Specifications

  • Sterility:
    Sterile
  • Description:
    Bovine calf serum
  • Origin:
    United States
  • Storage temperature:
    −40...−10°C
  • Pk:
    1 L

Specifications

About this item

Bovine calf serum, United States origin are collected at government approved abattoirs in the United States.

  • Cattle are less than 35 weeks of age at time of collection
  • True pooled and triple 0,1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
  • Shelf life: 60 months
  • Format: Liquid

Iron fortified to raise iron saturation to between 70 - 80% of total iron binding capacity.

Certifications: Granted a Certificate of Suitability, R0-CEP 2018-305, by the European Directorate for the Quality of Medicines (EDQM).