ContactUsLinkComponent
Reducer Fittings, Male NPT Threaded, Straight
Fittings, PFA, Straight, Threaded Reducing Nipples, 1/4" x 1/8" NPT(M); 1/Ea
Catalog # MFLX06546-23
undefined
Fittings, PFA, Straight, Threaded Reducing Nipples, 1/4" x 1/8" NPT(M); 1/Ea
Catalog # MFLX06546-23
Specifications
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    1/8"
  • Thread size NPT(M) (port 2):
    1/4"
  • Description:
    Fittings, PFA, Straight, Threaded Reducing Nipples, 1/4"to1/8" NPT(M); 1/Ea
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    1 year
  • Pk:
    1
  • Fitting material:
    PFA
Specifications
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings provide a more permanent fluid path. Use these fittings to reduce or enlarge connection sizes. Choose the size and material to meet your application needs.
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    1/8"
  • Thread size NPT(M) (port 2):
    1/4"
  • Description:
    Fittings, PFA, Straight, Threaded Reducing Nipples, 1/4"to1/8" NPT(M); 1/Ea
  • Environmentally Preferable:
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Warranty:
    1 year
  • Pk:
    1
  • Fitting material:
    PFA