Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, Nylon, Avantor®
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Environmentally Preferable:
- Catalog No:
- MFLX30624-25
- MFLX30624-23
- MFLX30624-19
- MFLX30624-21
- MFLX30401-64
- MFLX30624-15
- MFLX30624-13
- MFLX30624-05
- MFLX30624-02
- MFLX30624-11
- MFLX30628-13
- MFLX30624-07
- MFLX30623-85
- Model:
- 30624-25
- 30624-23
- 30624-19
- 30624-21
- 30401-64
- 30624-15
- 30624-13
- 30624-05
- 30624-02
- 30624-11
- 30628-13
- 30624-07
- 30623-85
- Orifice dimensions:
- 0,34375"
- 0,25"
- 0,5625"
- 0,125"
- 0,15625"
- 0,25"
- 0,125"
- 0,125"
- 0,25"
- 0,34375"
- 0,125"
- 0,25"
- 0,78125"
- Sterilisation:
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Warranty:
- 1 year
Frequently Bought Together