ContactUsLinkComponent
Masterflex® Union Fittings, Hose Barb, Straight, Stainless Steel, Avantor®
undefined
Masterflex® Union Fittings, Hose Barb, Straight, Stainless Steel, Avantor®
  • Model:
  • 30620-05
  • 30620-08
  • 31208-01
  • 30620-06
  • 31208-04
  • 30620-03
  • 30620-04
  • 30620-07
  • 31208-00
  • 31208-02
  • 30620-01
  • 30620-02
  • Orifice dimensions:
  • 7,938 mm (0,3125")
  • 17,46 mm (0,6875")
  • 1,588 mm (0,0625")
  • 9,53 mm (0,375")
  • 3,175 mm (0,125")
  • 4,763 mm (0,1875")
  • 1,588 mm (0,0625")
  • 1,588 mm (0,0625")
  • 0,7925 mm (0,03125")
  • 3,175 mm (0,125")
  • Sterilisation:
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
  • 1 Month
  • 1 Month
  • 1 year
  • 1 Month
  • 1 year
  • 1 Month
  • 1 Month
  • 1 Month
  • 1 year
  • 1 year
  • 1 Month
Frequently Bought Together