ContactUsLinkComponent
Masterflex® Quick-Disconnect Fittings, Avantor®
Masterflex® Quick-Disconnect Fittings, Avantor®
Certificates
undefined
Masterflex® Quick-Disconnect Fittings, Avantor®
Certificates
Specifications
  • Model:
  • 70000-37
  • 70000-27
  • 70000-38
  • Fitting depth:
  • 14 mm (0,5625")
  • Max. operating pressure:
  • 120 psi
  • 120 psi
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • O-Ring material:
  • Buna N
  • Buna-N
  • Valve material:
  • Acetal
  • Acetal (POM)
  • Valve spring material:
  • 316 SS
  • 316 SS
  • Warranty:
  • 1 year
  • 1 year