Masterflex® Reducer Fittings, Hose Barb, Straight (Polypropylene), Avantor®
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range 9 to 180 °F (-13 to +66 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Environmentally Preferable:
- Catalog No:
- MFLX40610-07
- MFLX40610-10
- MFLX40610-11
- MFLX40610-08
- MFLX40610-09
- MFLX40610-12
- MFLX50626-21
- MFLX50626-23
- MFLX30705-12
- MFLX40610-17
- MFLX40626-26
- MFLX40610-18
- MFLX40626-32
- MFLX40610-15
- MFLX40616-43
- MFLX40610-16
- MFLX40616-47
- MFLX40610-22
- MFLX50625-95
- MFLX40610-19
- MFLX40640-21
- MFLX40610-20
- MFLX40920-23
- MFLX40610-21
- MFLX40640-17
- MFLX40640-16
- MFLX40640-15
- MFLX40640-11
- MFLX40610-13
- MFLX40610-14
- MFLX40640-08
- MFLX40640-09
- MFLX40640-13
- MFLX40640-18
- MFLX40640-10
- MFLX41518-34
- MFLX40640-12
- MFLX41518-36
- MFLX30705-11
- MFLX40640-20
- MFLX30616-75
- MFLX30616-71
- MFLX50622-23
- MFLX50621-95
- MFLX50622-21
- MFLX40622-26
- MFLX40622-32
- MFLX40622-19
- Model:
- 40610-07
- 40610-10
- 40610-11
- 40610-08
- 40610-09
- 40610-12
- 50626-21
- 50626-23
- 30705-12
- 40610-17
- 40626-26
- 40610-18
- 40626-32
- 40610-15
- 40616-43
- 40610-16
- 40616-47
- 40610-22
- 50625-95
- 40610-19
- 40640-21
- 40610-20
- 40920-23
- 40610-21
- 40640-17
- 40640-16
- 40640-15
- 40640-11
- 40610-13
- 40610-14
- 40640-08
- 40640-09
- 40640-13
- 40640-18
- 40640-10
- 41518-34
- 40640-12
- 41518-36
- 30705-11
- 40640-20
- 30616-75
- 30616-71
- 50622-23
- 50621-95
- 50622-21
- 40622-26
- 40622-32
- 40622-19
- Orifice dimensions:
- 3,175 mm (0,125")
- 3,175 mm (0,125")
- 4,763 mm (0,1875")
- 3,175 mm (0,125")
- 4,763 mm (0,1875")
- 6,35 mm (0,25")
- 6,35 mm (0,25")
- 8,73 mm (0,34375")
- 6,35 mm (0,25")
- 8,73 mm (0,34375")
- 15,08 mm (0,59375")
- 11,9 mm (0,46875")
- 11,9 mm (0,46875")
- 8,73 mm (0,34375")
- 11,9 mm (0,46875")
- 6,35 mm (0,25")
- 8,73 mm (0,34375")
- 6,35 mm (0,25")
- 4,763 mm (0,1875")
- 3,175 mm (0,125")
- 4,763 mm (0,1875")
- 3,175 mm (0,125")
- 4,763 mm (0,1875")
- 3,175 mm (0,125")
- 8,73 mm (0,34375")
- 3,175 mm (0,125")
- 6,35 mm (0,25")
- 8,73 mm (0,34375")
- Sterilisation:
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene Oxide (ETO) or Autoclave
- Ethylene Oxide (ETO) or Gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene Oxide (ETO) or Autoclave
- Ethylene oxide (ETO) or gamma
- Ethylene Oxide (ETO) or Autoclave
- Ethylene oxide (ETO) or gamma
- Ethylene Oxide (ETO) or Autoclave
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Ethylene Oxide (ETO) or Gamma
- Ethylene Oxide (ETO) or Autoclave
- Ethylene Oxide and autoclave
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Ethylene oxide (ETO) or gamma
- Warranty:
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- 1 year
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- 1 year
- Out of box
Frequently Bought Together