ContactUsLinkComponent
Masterflex® Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, Avantor®
Masterflex® Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, Avantor®
Certificates
undefined
Masterflex® Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, Avantor®
Certificates
Specifications
  • Model:
  • 96124-18
  • 96124-05
  • 96124-11
  • 96124-16
  • Length per pack:
  • 7,6 m (25 ft.)
  • 7,6 m (25 ft.)
  • 7,6 m (25 ft.)
  • 7,6 m (25 ft.)
  • Max. pressure:
  • 17 psi
  • 35 psi
  • 28 psi
  • 23 psi
  • Max. temperature:
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • Min. temperature:
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • Regulatory approvals:
  • USP
  • USP
  • USP
  • USP
  • Sterilisation:
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Autoclave, Ethylene Oxide, Gamma Irradiation
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • Yes
  • Yes
  • Yes
  • Yes