ContactUsLinkComponent
Bürkle Thread Adapters for Hand-Operated Mini Solvent Pump
Burkle 0006-4002 Threaded Adapter for Mini Solvent Pump, ETFE; GL45-GL38
Catalog # MFLX06040-02
undefined
Burkle 0006-4002 Threaded Adapter for Mini Solvent Pump, ETFE; GL45-GL38
Catalog # MFLX06040-02
Specifications
  • Description:
    Threaded Adapter for Mini Solvent Hand Pump, ETFE; GL45-GL38
  • Environmentally Preferable:
  • Wetted materials:
    ETFE
  • Warranty:
    15 Months
  • Pk:
    1
  • Container adapter:
    GL38
Specifications
About this item
Suitable for use with Hand-Operated Mini Solvent Pumps.
  • Order optional adapters to fit a wide range of containers
  • Avaiable in different material and thread configurations
  • Withstand temperatures up to 104 °F (40 °C)
The 06043-95 hand pump features a GL45 connecting thread made of PTFE for connection to a wide range of containers. Optional S40, GL38 and GL32 adapters in PTFE or ETFE are available separately to easily adapt this hand operating pump to other bottles, canisters, or tanks.
  • Description:
    Threaded Adapter for Mini Solvent Hand Pump, ETFE; GL45-GL38
  • Environmentally Preferable:
  • Wetted materials:
    ETFE
  • Warranty:
    15 Months
  • Pk:
    1
  • Container adapter:
    GL38