ContactUsLinkComponent
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PCS, Double-Y Elements, Avantor®
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PCS, Double-Y Elements, Avantor®
Certificates
undefined
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PCS, Double-Y Elements, Avantor®
Certificates
Specifications
  • Clarity:
  • Opaque
  • Opaque
  • GMP compliant:
  • Yes
  • Yes
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Max. temperature:
  • 150 °C (302 °F)
  • 150 °C (302 °F)
  • Pump series:
  • Ismatec
  • Ismatec
  • Regulatory approvals:
  • ADCF;RoHS;USP
  • ADCF;RoHS;USP
  • Series:
  • Ismatec
  • Ismatec
  • Sterilisation:
  • Autoclave
  • Autoclave
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Sterilisation method EtO:
  • No
  • No
  • Sterilisation method gamma:
  • No
  • No
  • Warranty:
  • Out of box
  • Out of box