ContactUsLinkComponent
PrimeLock® Benchtop Installation Tool, 1/4" to 1" OD tubing
  MFLX30493-25
2 month warranty
PrimeLock® Benchtop Installation Tool, 1/4" to 1" OD tubing
  MFLX30493-25
2 month warranty
  • Description:
    Benchtop Installation Tool, 1/4" to 1" OD tubing
  • Compatible with:
    PrimeLock High-Purity PFA Flare Fittings, 1/4 to 1" OD Tubing
  • Pk:
    1
  • Warranty:
    2 months
Easily install your High-Purity PFA Fittings.
  • Optimize performance of your PFA PrimeLock® Fittings
Handheld and benchtop installation tools help you to quickly and easily install the PrimeLock insert correctly every time. Benchtop tool is the quickest and easiest method of installing the insert when space constraints are not an issue. The compact, handheld tool enables you to install the insert in remote locations. Installation tools require mandrel(s) and clamp insert(s) for operation.
  • Description:
    Benchtop Installation Tool, 1/4" to 1" OD tubing
  • Environmentally Preferable:
  • Compatible with:
    PrimeLock High-Purity PFA Flare Fittings, 1/4 to 1" OD Tubing
  • Pk:
    1
  • Warranty:
    2 months