ContactUsLinkComponent
LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
Certificates
undefined
LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
Specifications
  • Accuracy:
  • ±3%
  • ±3%
  • Max. pressure:
  • 94 psi
  • 94 psi
  • Min. operating temperature:
  • 10 °C (50 °F)
  • 10 °C (50 °F)
  • Max. operating temperature:
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • Viscosity:
  • 0,7 to 10 cSt
  • 0,7 to 10 cSt
  • Wetted materials:
  • None
  • None
  • Warranty:
  • 1 year
  • 1 year