About this item
Capable of speeds up to 16,100 ×g.
- Save bench space with 9" wide centrifuge
- Quick deceleration for shorter down time
- Can hold up to 12 x 1.5/2.0 ml tubes
- Control knob and bright LCD provide clear indication of time and speed
This compact microcentrifuge offers speed and force usually found on larger models. The unit delivers shortened acceleration and deceleration time with a quiet motor. The brushless, maintenance free drive system provides a powerful, smooth operation. A separate momentary spin button permits instant spin downs, without setting or disturbing any preset parameters.
- Environmentally Preferable:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are: