ContactUsLinkComponent
Masterflex® L/S® Cytoflow™ Pump Heads, Avantor®
undefined
Masterflex® L/S® Cytoflow™ Pump Heads, Avantor®
  • Model:
  • 77253-12
  • 77253-10
  • 77253-02
  • 77253-00
  • Gear ratio:
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • Occlusion:
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Open-head sensor:
  • No
  • No
  • No
  • No
  • Optimum RPM range:
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • Tubing retention:
  • Manual
  • Manual
  • Manual
  • Manual
  • Tubing sizes accepted:
  • L/S® 15, L/S 24, L/S 35, L/S 36
  • L/S® 16, L/S 25, L/S 17, L/S 18
  • L/S® 15, L/S 24, L/S 35, L/S 36
  • L/S® 16, L/S 25, L/S 17, L/S 18
  • Volume per revolution with L/S 15 tubing:
  • 3,5 ml
  • 4 ml
  • Volume per revolution with L/S 16 tubing:
  • 1,6 ml
  • 1,7 ml
  • Volume per revolution with L/S 17 tubing:
  • 5,6 ml
  • 6,1 ml
  • Volume per revolution with L/S 18 tubing:
  • 7,7 ml
  • 8,6 ml
  • Volume per revolution with L/S 24 tubing:
  • 5,7 ml
  • 6,2 ml
  • Volume per revolution with L/S 25 tubing:
  • 3,5 ml
  • 3,9 ml
  • Volume per revolution with L/S 35 tubing:
  • 7,8 ml
  • 9,5 ml
  • Volume per revolution with L/S 36 tubing:
  • 9,6 ml
  • 12,8 ml
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Frequently Bought Together