ContactUsLinkComponent
Masterflex® L/S® High-Performance Pump Heads, Avantor®
Masterflex® L/S® High-Performance Pump Heads, Avantor®
Certificates
undefined
Masterflex® L/S® High-Performance Pump Heads, Avantor®
Specifications
  • Model:
  • 77252-80
  • 77252-72
  • 77252-70
  • 77252-62
  • 77252-60
  • 77252-82
  • Gear ratio:
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • Occlusion:
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Open-head sensor:
  • No
  • No
  • No
  • Yes
  • Yes
  • No
  • Optimum RPM range:
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • Suction lift:
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • Tubing retention:
  • Manual
  • Manual
  • Manual
  • Manual
  • Manual
  • Manual
  • Tubing sizes accepted:
  • L/S® 16, L/S 25, L/S 17, L/S 18
  • L/S® 15, L/S 24, L/S 35, L/S 36, L/S 14HP, L/S 16HP, L/S 15HP, L/S 24HP
  • L/S® 16, L/S 25, L/S 17, L/S 18
  • L/S® 15, L/S 24, L/S 35, L/S 36, L/S 14HP, L/S 16HP, L/S 15HP, L/S 24HP
  • L/S® 16, L/S 25, L/S 17, L/S 18
  • L/S® 15, L/S 24, L/S 35, L/S 36, L/S 14HP, L/S 16HP, L/S 15HP, L/S 24HP
  • Volume per revolution with L/S 15 tubing:
  • 1,8 ml
  • 1,8 ml
  • 1,8 ml
  • Volume per revolution with L/S 16 tubing:
  • 0,9 ml
  • 0,9 ml
  • 0,9 ml
  • Volume per revolution with L/S 17 tubing:
  • 3 ml
  • 3 ml
  • 3 ml
  • Volume per revolution with L/S 18 tubing:
  • 3,8 ml
  • 3,8 ml
  • 3,8 ml
  • Volume per revolution with L/S 24 tubing:
  • 3 ml
  • 3 ml
  • 3 ml
  • Volume per revolution with L/S 25 tubing:
  • 1,9 ml
  • 1,9 ml
  • 1,9 ml
  • Volume per revolution with L/S 35 tubing:
  • 4,3 ml
  • 4,3 ml
  • 4,3 ml
  • Volume per revolution with L/S 36 tubing:
  • 5,8 ml
  • 5,8 ml
  • 5,8 ml
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years