ContactUsLinkComponent
Value Plastics ABSML-6005-1 Fitting, White Nylon over Polypropylene, Male Luer with Rotating Lock Ring, 5/16" Hex to 1/4-28 UNF Thread, 1000/PK
Catalog # MFLX15973-45BLK
undefined
Value Plastics ABSML-6005-1 Fitting, White Nylon over Polypropylene, Male Luer with Rotating Lock Ring, 5/16" Hex to 1/4-28 UNF Thread, 1000/PK
Catalog # MFLX15973-45BLK
Specifications
  • Connection Type:
    Male Luer to Thread
  • Flow pattern:
    Straight
  • Description:
    ABSML-6005-1 Fitting, White Nylon over Polypropylene, Male Luer with Rotating Lock Ring, ⁵⁄₁₆" Hex to 1/4-28 UNF Thread, 1000/PK
  • Connection type (port 1):
    Male Luer with Rotating Lock Ring
  • No. of ports:
    2
  • Warranty:
    None
  • Pk:
    1.000
  • Fitting material:
    Nylon
  • Thread size UNF(M) (port 1):
    ¼"-28
Specifications
About this item
Premium luer fittings for critical applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Manufactured from 100% virgin raw materials in a Class 100000 environment
  • Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options. All fittings are sold in packs of 100.
  • Connection Type:
    Male Luer to Thread
  • Flow pattern:
    Straight
  • Description:
    ABSML-6005-1 Fitting, White Nylon over Polypropylene, Male Luer with Rotating Lock Ring, ⁵⁄₁₆" Hex to 1/4-28 UNF Thread, 1000/PK
  • Environmentally Preferable:
  • Connection type (port 1):
    Male Luer with Rotating Lock Ring
  • No. of ports:
    2
  • Warranty:
    None
  • Pk:
    1.000
  • Fitting material:
    Nylon
  • Thread size UNF(M) (port 1):
    ¼"-28