ContactUsLinkComponent
Aalborg® Paddle Wheel Flowmeters for Monitoring and Control
Aalborg® Paddle Wheel Flowmeters for Monitoring and Control
Certificates
undefined
Aalborg® Paddle Wheel Flowmeters for Monitoring and Control
Specifications
  • Model:
  • 32755-44
  • 32755-35
  • 32755-46
  • 32755-36
  • 32755-38
  • 32755-45
  • 32755-31
  • 32755-34
  • 32755-32
  • 32755-24
  • 32755-18
  • 32755-17
  • Accuracy:
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • ±1% full-scale
  • Communication interface:
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • RS-232
  • Connection:
  • 12-pin M16 shielded cable
  • 12-pin M16 shielded cable
  • 12-pin M16 shielded cable
  • 12-pin M16 shielded cable
  • 12-pin M16 shielded cable
  • 12-pin M16 shielded cable
  • 12-pin M16 shielded cable
  • 12-pin M16 shielded cable
  • 12-pin M16 shielded cable
  • 3-wire 6-ft shielded cable
  • 3-wire 6-ft shielded cable
  • 3-wire 6-ft shielded cable
  • Display type:
  • 2-line x 16-character LCD, 0,5"H
  • 2-line x 16-character LCD, 0,5"H
  • 2-line x 16-character LCD, 0,5"H
  • 2-line x 16-character LCD, 0,5"H
  • 2-line x 16-character LCD, 0,5"H
  • 2-line x 16-character LCD, 0,5"H
  • 2-line x 16-character LCD, 0,5"H
  • 2-line x 16-character LCD, 0,5"H
  • 2-line x 16-character LCD, 0,5"H
  • Max. operating temperature:
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • O-Ring material:
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • EPDM
  • EPDM
  • EPDM
  • Pressure drop at max. flow:
  • 15 psid
  • 20 psid
  • 20 psid
  • 20 psid
  • Wetted materials:
  • PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
  • PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
  • PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
  • PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
  • PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
  • PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
  • PVDF, zirconia ceramic, saphhire, PTFE, and 316 SS
  • PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
  • PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
  • PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
  • PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
  • PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year