ContactUsLinkComponent
Remote-Control Diaphragm Liquid Dosing Pumps
Remote-Control Diaphragm Liquid Dosing Pumps
Certificates
undefined
Remote-Control Diaphragm Liquid Dosing Pumps
Specifications
  • Inlet connection:
  • 5/16" hose barb
  • 5/16" hose barb
  • Max. pressure:
  • 5,86 bar (85 psi)
  • 5,86 bar (85 psi)
  • Max. temperature:
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • Outlet connection:
  • 5/16" hose barb
  • 5/16" hose barb
  • Power cord:
  • 6-ft power cord with 3-prong U.S. plug
  • 6-ft power cord with 3-prong U.S. plug
  • Run dry:
  • Yes
  • Yes
  • Suction lift, wet:
  • 3962 mm of H₂O (156" H₂O)
  • 3962 mm of H₂O (156" H₂O)
  • Wetted materials:
  • PVDF head, PTFE diaphragm, FFKM valves
  • PVDF head, PTFE diaphragm, FFKM valves
  • Warranty:
  • 1 year
  • 1 year