ContactUsLinkComponent
Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Avantor®
Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Avantor®
Certificates
undefined
Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Avantor®
Certificates
Specifications
  • Clarity:
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Length per pack:
  • 30,5 m (100 ft.)
  • 30,5 m (100 ft.)
  • 30,5 m (100 ft.)
  • 30,5 m (100 ft.)
  • 30,5 m (100 ft.)
  • Max. temperature:
  • 85 °C (185 °F)
  • 85 °C (185 °F)
  • 85 °C (185 °F)
  • 85 °C (185 °F)
  • 85 °C (185 °F)
  • Min. temperature:
  • –31 °C (–25 °F)
  • –31 °C (–25 °F)
  • –31 °C (–25 °F)
  • –31 °C (–25 °F)
  • –31 °C (–25 °F)
  • Regulatory approvals:
  • Non-DEHP;USP
  • Non-DEHP;USP
  • Non-DEHP;USP
  • Non-DEHP;USP
  • Non-DEHP;USP
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes