ContactUsLinkComponent
DC-Powered Mini Diaphragm Pumps
undefined
DC-Powered Mini Diaphragm Pumps
  • Accuracy:
  • ±3%
  • ±3%
  • ±3%
  • ±3%
  • ±3%
  • Duty cycle intermittent:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • Input:
  • 0-5 V Proportional
  • Pulse
  • 0-5 V Proportional
  • 4-20 mA Proportional
  • 4-20 mA Proportional
  • Max. pressure:
  • 29 bar (2 psi)
  • 29 bar (2 psi)
  • 29 bar (2 psi)
  • 29 bar (2 psi)
  • 29 bar (2 psi)
  • Max. temperature:
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Max. viscosity:
  • 50 cP
  • 50 cP
  • 50 cP
  • 50 cP
  • 50 cP
  • Min. flow rate:
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • 0,055 ml/min (0,001 gal./h; 0 strokes/min)
  • Outlet connection:
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • 1/8" ID x 1/4" OD compression
  • Wetted materials:
  • Ceramic, FKM, PTFE, PVDF
  • Ceramic, FKM, PTFE, PVDF
  • Ceramic, FKM, PTFE, PVDF
  • Ceramic, FKM, PTFE, PVDF
  • Ceramic, FKM, PTFE, PVDF
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together