ContactUsLinkComponent
CPC® AseptiQuik® S Series Genderless Aseptic Connectors, Low Flow
CPC® AseptiQuik® S Series Genderless Aseptic Connectors, Low Flow
Certificates
undefined
CPC® AseptiQuik® S Series Genderless Aseptic Connectors, Low Flow
Specifications
  • Max. operating pressure:
  • 60 psi
  • 60 psi
  • 60 psi
  • 60 psi
  • 60 psi
  • 60 psi
  • Max. temperature:
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Min. temperature:
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • Seal material:
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Platinim-cured silicone
  • Sterilisation:
  • Autoclave or gamma
  • Gamma
  • Autoclave or gamma
  • Gamma
  • Autoclave or gamma
  • Gamma