Masterflex® Centrifuge Tube Adapters, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Use tube adapters in centrifuge rotors and buckets to accommodate your tube size.
- Environmentally Preferable:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.