Orders LinkContactUsLinkComponent
Precision Digital ProtEX Explosion-Proof Process Meters
Precision Digital ProtEX Explosion-Proof Process Meters
Process meter, explosion-proof, input loop powered with backlight
Catalog # MFLX30004-68
CE Compliant
Precision Digital ProtEX Explosion-Proof Process Meters
Catalog # MFLX30004-68
CE Compliant
Precision Digital ProtEX Explosion-Proof Process Meters
Process meter, explosion-proof, input loop powered with backlight
Price
992,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Four sensors operate as through-glass buttons for easy on-site programming without removing the cover and exposing the electronics.
  • High-contrast backlit LCD is easy to read from a distance and under most lighting conditions
  • 5-digit rate and 7-character total/tag display
  • 32-point, square root, or exponential linearization
  • 90° rotatable display
  • 5-digit password prevents unauthorized changes to the programmed parameter settings
  • Explosion-proof, IP68, NEMA 4X enclosure
  • FM, CSA, ATEX, IECEx, CE approved
The ProtEX PD6800 is a rugged, explosion-proof process meter designed for demanding applications in hazardous areas or in harsh environmental conditions. The meter derives all of its power from the 4 to 20 mA loop. Easily program using the four SafeTouch™ through-glass buttons without having to remove the cover or use the four mechanical buttons. Unit features backlit display, min/max readings, password protection, and alarm indication. Die-cast aluminum enclosure has two ¾" threaded conduit holes and integrated pipe or wall mounting flanges.
Specifications
  • Description:
    Process meter, explosion-proof, input loop powered with backlight
  • Cat. No.:
    MFLX30004-68
  • Warranty:
    3 years