ContactUsLinkComponent
Masterflex® Transfer Tubing, Cleargreen® Phthalate and DEHP-Free PVC, Avantor®
Masterflex® Transfer Tubing, Cleargreen® Phthalate and DEHP-Free PVC, Avantor®
Certificates
undefined
Masterflex® Transfer Tubing, Cleargreen® Phthalate and DEHP-Free PVC, Avantor®
Specifications
  • Clarity:
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Length per pack:
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • 15,2 m (50 ft.)
  • Max. temperature:
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • 73 °C (165 °F)
  • Min. temperature:
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • –50 °C (–58 °F)
  • Regulatory approvals:
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • ADCF;FDA;Non-DEHP;Non-Phthalate;USP
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No