ContactUsLinkComponent
Masterflex® FDA Compliant Sanitary Drum Pumps, Avantor®
Masterflex® FDA Compliant Sanitary Drum Pumps, Avantor®
Certificates
undefined
Masterflex® FDA Compliant Sanitary Drum Pumps, Avantor®
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Model:
  • 75219-15
  • 75219-55
  • 75219-60
  • 75219-05
  • 75219-20
  • Air consumption:
  • 28 ft.³/min
  • 28 ft.³/min
  • CE regulatory approval:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 1 5/8"
  • 1 5/8"
  • 1 5/8"
  • 1 5/8"
  • 1 5/8"
  • Max. air inlet pressure:
  • 6,2 bar (90 psig)
  • Max. temperature:
  • 80 °C (175 °F)
  • 80 °C (175 °F)
  • 80 °C (175 °F)
  • 80 °C (175 °F)
  • 80 °C (175 °F)
  • Max. viscosity:
  • 1000 cP
  • 1000 cP
  • 1000 cP
  • 1000 cP
  • 1000 cP
  • Motor type:
  • TEFC
  • TEFC
  • Air
  • ODP
  • Air
  • Outlet connection:
  • 1 1/4" Tri-Clamp
  • 1 1/4" Tri-Clamp
  • 1 1/4" Tri-Clamp
  • 1 1/4" Tri-Clamp
  • 1 1/4" Tri-Clamp
  • Tube length, inlet:
  • 39"
  • 39"
  • 39"
  • 39"
  • 39"
  • Wetted materials:
  • 316SS, BUNA N, PTFE
  • 316SS, BUNA N, PTFE
  • 316SS, BUNA N, PTFE
  • 316SS, BUNA N, PTFE
  • 316SS, BUNA N, PTFE
  • Warranty:
  • 3 years
  • 3 years
  • 3 years
  • 3 years
  • 3 years