ContactUsLinkComponent
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories
Certificates
undefined
PendoTECH® Single-Use Pressure Sensor Accessories
Certificates
Specifications
  • Max. pressure:
  • 76 psi
  • 76 psi
  • 76 psi
  • Process connection:
  • 1/4" x 1/4" hose barb, male luer
  • 3/8" x 3/8" hose barb, male luer
  • 1/2" x 1/2" hose barb, male luer
  • Length:
  • 304,8 cm (120")
  • 182,88 cm (72")
  • 365,76 cm (144")
  • 213,36 cm (84")
  • 365,76 cm (144")
  • 182,88 cm (72")
  • Warranty:
  • 2 years
  • 2 years
  • 3 Months
  • 1 year
  • 2 years
  • 1 year
  • 2 years
  • 2 years
  • 2 years
  • 3 Months
  • 2 years
  • 2 years
  • 2 years
  • 3 Months
  • 2 years
  • 2 years