Orders LinkContactUsLinkComponent
Masterflex® DB-9 Pump Interface Cables, Avantor®
Masterflex® DB-9 Pump Interface Cables, Avantor®
Interface cable with DB9 male/DB9 female connectors
Catalog # MFLX22050-54
Masterflex® DB-9 Pump Interface Cables, Avantor®
Catalog # MFLX22050-54
Masterflex® DB-9 Pump Interface Cables, Avantor®
Masterflex® DB-9 Pump Interface Cables, Avantor®
Interface cable with DB9 male/DB9 female connectors
Price
27,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
Optimize performance of your Masterflex® pump.
  • Suitable for use with Masterflex® L/S® drives with a 9-pin connector
Use these 9-pin interface cable assemblies with Masterflex L/S drives having a 9-pin connector; including the 07550- and 07551-series computer-compatible drives. Cable measures 6 ft (1.8 m) long.
Specifications
  • Description:
    Interface cable with DB9 male/DB9 female connectors
  • Cat. No.:
    MFLX22050-54
  • Warranty:
    LTD life time
  • Model:
    22050-54
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • USB to DB9 (M) RS-232 serial adapter cable
    USB to DB9 (M) RS-232 serial adapter cable
    MFLX22050-58
    Log in for availability
    1 * 1 ST - 65,90 €