Orders LinkContactUsLinkComponent
Flowline DataView Advanced Meter & Controller
Flowline DataView Advanced Meter & Controller
Flowline DataView Advanced Meter & Controller
Certificates
Flowline DataView Advanced Meter & Controller
3 Options Available Below
Product Details & Documents
Easy configuration via push button display module.
  • Bright 6-digit LED main display and secondary unit display with relay status indicators
  • 1/8 DIN enclosure with NEMA 4X faceplate and shallow depth case
  • 32-point linearization function for volumetric tank measurement
  • Non-volatile memory with security password protected lock out
  • Isolated 4-20 mA repeater output with 40 mA internal power supply
The general purpose meter displays engineering units with one 4-20 mA or 0-10 VDC transmitter input channel and is offered in three configurations with optional 2 or 4 relays, 4 or 8 set points and an isolated 4-20 mA repeater. Each relay can be configured on a single set point or latched on two set points for automatic fill or empty control including duplexing. For field mount installations,
order the single or two meter NEMA box.
Specifications for Product Family
Specifications
  • Signal output:
  • 4 to 20 mA
  • 4 to 20 mA
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • Power
    Availability
    Price
  • MFLX23002-18
    Advanced Meter, No Outputs; 24 VDC
    Advanced Meter, No Outputs; 24 VDC
    85 - 265 VAC; 24 VDC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 340,00 €
    1 340,00 €
    1 * 1 ST
       1 340,00 €
    Specifications:
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX23002-18
    Signal output
    Warranty
    1 year
    IP65 washdown
  • MFLX23002-20
    Advanced Meter/Controller, 2 Relays; 24 VDC
    Advanced Meter/Controller, 2 Relays; 24 VDC
    85 - 265 VAC; 24 VDC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 550,00 €
    1 550,00 €
    1 * 1 ST
       1 550,00 €
    Specifications:
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX23002-20
    Signal output
    4 to 20 mA
    Warranty
    1 year
    IP65 washdown
  • MFLX23002-22
    Advanced Meter/Controller, 4 Relays; 24 VDC
    Advanced Meter/Controller, 4 Relays; 24 VDC
    85 - 265 VAC; 24 VDC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 710,00 €
    1 710,00 €
    1 * 1 ST
       1 710,00 €
    Specifications:
    IP65 washdown
    More
    Specifications
    Cat. No.
    MFLX23002-22
    Signal output
    4 to 20 mA
    Warranty
    1 year
    IP65 washdown