Orders LinkContactUsLinkComponent
CPC® Quick-Disconnect Fittings, Threaded Inserts
CPC® Quick-Disconnect Fittings, Threaded Inserts
CPC® Quick-Disconnect Fittings, Threaded Inserts
Certificates
CPC® Quick-Disconnect Fittings, Threaded Inserts
CPC® Quick-Disconnect Fittings, Threaded Inserts
10 Options Available Below
Product Details & Documents
No more struggling - just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
Documents
Specifications for Product Family
Specifications
  • Max. operating pressure:
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • 120 psi
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • 82 °C (180 °F)
  • 71 °C (160 °F)
  • 82 °C (180 °F)
  • 71 °C (160 °F)
  • Max. vacuum:
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • 711 mm Hg (28" Hg)
  • Seal material:
  • Buna N
  • Buna N
  • Buna N
  • Buna N
  • EPDM
  • EPDM
  • Buna N
  • EPDM
  • Buna N
  • EPDM
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Gamma
  • Gamma
  • Autoclave or ethylene oxide (ETO)
  • Gamma
  • Autoclave or ethylene oxide (ETO)
  • Gamma
  • Valve spring material:
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Bulk product
Product Information
  • Type
    Connection Type
    Fitting material
    Flow pattern
    Quick-disconnect flow size
    Thread size NPT(M) (port 1)
    Thread size NPT(M) (port 2)
    Supplier No.
    Availability
    Price
  • MFLX06361-29BLK
    Quick-Disconnect Fitting, Threaded Insert, Acetal, Valved, ¹/₈" ID, PMCD2402; 25/PK
    Quick-Disconnect Fitting, Threaded Insert, Acetal, Valved, ¹/₈" ID, PMCD2402; 25/PK
    Valved
    Quick-Disconnect to Thread
    Acetal
    Straight
    ⅛"
    ⅛"
    ⅛"
    PMCD2402 (25PK)
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 212,00 €
    212,00 €
    1 * 25 ST
       212,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-29BLK
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06361-31BLK
    Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₈" Flow Size; ¹/₄" ID, PMCD2404; 25/PK
    Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₈" Flow Size; ¹/₄" ID, PMCD2404; 25/PK
    Valved
    Quick-Disconnect to Thread
    Acetal
    Straight
    ⅛"
    ¼"
    ⅛"
    PMCD2404 (25PK)
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 217,00 €
    217,00 €
    1 * 25 ST
       217,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-31BLK
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06361-81BLK
    Quick-Disconnect Fitting, Thread Insert, Polypropylene, Straight-Through; ¹/₄" ID, PLC2400412; 25/PK
    Quick-Disconnect Fitting, Thread Insert, Polypropylene, Straight-Through; ¹/₄" ID, PLC2400412; 25/PK
    Quick-Disconnect to Thread
    Polypropylene
    Straight
    ¼"
    ¼"
    ¼"
    PLC2400412 (25PK)
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 186,00 €
    186,00 €
    1 * 25 ST
       186,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-81BLK
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    Warranty
    1 year
  • MFLX06361-65BLK
    Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₄" Flow Size; ¹/₄" ID, PLCD24004; 25/PK
    Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₄" Flow Size; ¹/₄" ID, PLCD24004; 25/PK
    Valved
    Quick-Disconnect to Thread
    Acetal
    Straight
    ¼"
    ¼"
    ¼"
    PLCD24004 (25PK)
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 204,00 €
    204,00 €
    1 * 25 ST
       204,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-65BLK
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06361-91BLK
    Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, ¹/₄" ID, PLCD2400412; 25/PK
    Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, ¹/₄" ID, PLCD2400412; 25/PK
    Valved
    Quick-Disconnect to Thread
    Polypropylene
    Straight
    ¼"
    ¼"
    ¼"
    PLCD2400412 (25PK)
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 317,00 €
    317,00 €
    1 * 25 ST
       317,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-91BLK
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
Down pack product
Product Information
  • Type
    Connection Type
    Fitting material
    Flow pattern
    Quick-disconnect flow size
    Thread size NPT(M) (port 1)
    Thread size NPT(M) (port 2)
    Supplier No.
    Availability
    Price
  • MFLX06361-29
    Quick-Disconnect Fitting, Threaded Insert, Acetal, Valved, ¹/₈" ID, PMCD2402; 1/PK
    Quick-Disconnect Fitting, Threaded Insert, Acetal, Valved, ¹/₈" ID, PMCD2402; 1/PK
    Valved
    Quick-Disconnect to Thread
    Acetal
    Straight
    ⅛"
    ⅛"
    ⅛"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,20 €
    14,20 €
    1 * 1 ST
       14,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-29
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06361-31
    Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₈" Flow Size; ¹/₄" ID, PMCD2404; 1/PK
    Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₈" Flow Size; ¹/₄" ID, PMCD2404; 1/PK
    Valved
    Quick-Disconnect to Thread
    Acetal
    Straight
    ⅛"
    ¼"
    ⅛"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,90 €
    13,90 €
    1 * 1 ST
       13,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-31
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06361-65
    Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₄" Flow Size; ¹/₄" ID, PLCD24004; 1/PK
    Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₄" Flow Size; ¹/₄" ID, PLCD24004; 1/PK
    Valved
    Quick-Disconnect to Thread
    Acetal
    Straight
    ¼"
    ¼"
    ¼"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,40 €
    14,40 €
    1 * 1 ST
       14,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-65
    Max. operating pressure
    120 psi
    Max. temperature
    82 °C (180 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    Buna N
    Sterilisation
    Autoclave or ethylene oxide (ETO)
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06361-91
    Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, ¹/₄" ID, PLCD2400412; 1/PK
    Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, ¹/₄" ID, PLCD2400412; 1/PK
    Valved
    Quick-Disconnect to Thread
    Polypropylene
    Straight
    ¼"
    ¼"
    ¼"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,50 €
    21,50 €
    1 * 1 ST
       21,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-91
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    316 Stainless Steel
    Warranty
    1 year
  • MFLX06361-81
    Quick-Disconnect Fitting, Thread Insert, Polypropylene, Straight-Through; ¹/₄" ID, PLC2400412; 1/PK
    Quick-Disconnect Fitting, Thread Insert, Polypropylene, Straight-Through; ¹/₄" ID, PLC2400412; 1/PK
    Quick-Disconnect to Thread
    Polypropylene
    Straight
    ¼"
    ¼"
    ¼"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,90 €
    11,90 €
    1 * 1 ST
       11,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX06361-81
    Max. operating pressure
    120 psi
    Max. temperature
    71 °C (160 °F)
    Max. vacuum
    711 mm Hg (28" Hg)
    Seal material
    EPDM
    Sterilisation
    Gamma
    Valve spring material
    Warranty
    1 year